6-Benzylaminopurin -Lösung 1,0 mg/ml

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzyl(purin-6-yl)amin, 6-Benzyladenin, N6-Benzyladenin

BIPB130-100ML BIPB130-500ML
APOSBIPB130-100MLEA 50.9 EUR
APOSBIPB130-100ML APOSBIPB130-500ML
6-Benzylaminopurin -Lösung 1,0 mg/ml
6-Benzylaminopurin

  • Plant tissue culture tested

Formel: C₁₂H₁₁N₅
Lagertemperatur: Kühlschrank
MDL: MFCD00005572
CAS-Nummer: 1214-39-7
EINECS: 214-927-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR