Ald-PEG4-t-butyl ester
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 1-oxo-3,6,9,12-tetraoxapentadecan-15-oate
BIPG1014-100MG
APOSBIPG1014-100MGEA
763
EUR
APOSBIPG1014-100MG
Ald-PEG4-t-butyl ester
Ald-PEG4-t-butyl ester
Formel:
C₁₅H₂₈O₇ Molecular Weight: 320,379 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25424117 CAS-Nummer: 1415329-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...