Ald-Ph-PEG5-t-butyl ester

Lieferant: Apollo Scientific

BIPG1026-100MG
APOSBIPG1026-100MGEA 493 EUR
APOSBIPG1026-100MG
Ald-Ph-PEG5-t-butyl ester
Ald-Ph-PEG5-t-butyl ester
Formel: C₂₂H₃₄O₈
Molecular Weight: 426,501 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24539463
CAS-Nummer: 1433996-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR