Amino-PEG3-acid

Lieferant: Apollo Scientific
Achtung

Synonyme: Amino-PEG3-C2-acid, 3-(2-(2-(2-Aminoethoxy)ethoxy)ethoxy)propanoic acid

BIPG1032-250MG
APOSBIPG1032-250MGEA 27.7 EUR
APOSBIPG1032-250MG
Amino-PEG3-acid
Amino-PEG3-acid
Formel: C₉H₁₉NO₅
Molecular Weight: 221,251 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21607085
CAS-Nummer: 784105-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR