Amino-PEG5-alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: NH2-PEG5-OH, 14-Amino-3,6,9,12-tetraoxatetradecan-1-ol

BIPG1047-100MG
APOSBIPG1047-100MGEA 40.6 EUR
APOSBIPG1047-100MG
Amino-PEG5-alcohol
Amino-PEG5-alcohol
Formel: C₁₀H₂₃NO₅
Molecular Weight: 237,293 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD28950782
CAS-Nummer: 34188-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR