1,14-Diamino-3,6,9,12-tetraoxatetradecane

Lieferant: Apollo Scientific
Achtung

Synonyme: Triethylene Glycol bis(2-aminoethyl) ether, 3,6,9,12-Tetraoxatetradecane-1,14-diamine

BIPG1056-250MG
APOSBIPG1056-250MGEA 461 EUR
APOSBIPG1056-250MG
1,14-Diamino-3,6,9,12-tetraoxatetradecane
1,14-Diamino-3,6,9,12-tetraoxatetradecane
Formel: C₁₀H₂₄N₂O₄
Molecular Weight: 236,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01365938
CAS-Nummer: 68960-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR