Azido-PEG6-Acid

Lieferant: Apollo Scientific
Achtung

BIPG1108-100MG BIPG1108-50MG BIPG1108-250MG
APOSBIPG1108-100MGEA 0 EUR
APOSBIPG1108-100MG APOSBIPG1108-50MG APOSBIPG1108-250MG
Azido-PEG6-Acid
Azido-PEG6-Acid
Formel: C₁₅H₂₉N₃O₈
Lagertemperatur: Kühlschrank
MDL: MFCD22056306
CAS-Nummer: 361189-66-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR