Azido-PEG6-Acid
Lieferant: Apollo Scientific
|
Achtung
|
BIPG1108-100MG
BIPG1108-50MG
BIPG1108-250MG
APOSBIPG1108-100MGEA
0
EUR
APOSBIPG1108-100MG
APOSBIPG1108-50MG
APOSBIPG1108-250MG
Azido-PEG6-Acid
Azido-PEG6-Acid
Formel:
C₁₅H₂₉N₃O₈ Lagertemperatur: Kühlschrank |
MDL:
MFCD22056306 CAS-Nummer: 361189-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...