Azido-PEG10-acid

Lieferant: Apollo Scientific
Achtung

BIPG1111-100MG
APOSBIPG1111-100MGEA 499 EUR
APOSBIPG1111-100MG
Azido-PEG10-acid
Azido-PEG10-acid
Formel: C₂₃H₄₅N₃O₁₂
Lagertemperatur: Kühlschrank
MDL: MFCD26127811
CAS-Nummer: 1644163-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR