Azido-PEG4-Amine 97%
Lieferant: Apollo Scientific
Synonyme:
Amino-PEG4-Azide, 14-Amino-3,6,9,12-tetraoxatetradecanyl Azide, 14-Azido-3,6,9,12-tetraoxatetradecan-1-amine, 1-Amino-14-azido-3,6,9,12-tetraoxatetradecane
BIPG1127-1G
APOSBIPG1127-1GEA
393
EUR
APOSBIPG1127-1G
Azido-PEG4-Amine 97%
Azido-PEG4-Amine
Formel:
C₁₀H₂₂N₄O₄ Molecular Weight: 262,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20134134 CAS-Nummer: 951671-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...