Azido-PEG6-amine
Lieferant: Apollo Scientific
Synonyme:
Azido-PEG-amine(n=6), O-(2-Aminoethyl)-O′-(2-azidoethyl)pentaethylene glycol, Azido-PEG6-amine, N3-PEG6-CH2CH2NH2
BIPG1129-1G
APOSBIPG1129-1GEA
654
EUR
APOSBIPG1129-1G
Azido-PEG6-amine
Azido-PEG6-amine
Formel:
C₁₄H₃₀N₄O₆ Molecular Weight: 350,411 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03453240 CAS-Nummer: 957486-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...