Azido-PEG8-amine

Lieferant: Apollo Scientific
Achtung

BIPG1131-1G
APOSBIPG1131-1GEA 763 EUR
APOSBIPG1131-1G
Azido-PEG8-amine
Azido-PEG8-amine
Formel: C₁₈H₃₈N₄O₈
Molecular Weight: 438,516 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03453241
CAS-Nummer: 857891-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR