Azido-PEG10-amine

Lieferant: Apollo Scientific

Synonyme: N3-PEG10-CH2CH2NH2, Azido-PEG-amine (n=10), 32-Azido-3,6,9,12,15,18,21,24,27,30-decaoxadotriacontan-1-amine, O-(2-Aminoethyl)-O′-(2-azidoethyl)nonaethylene glycol

BIPG1132-1G
APOSBIPG1132-1GEA 1010 EUR
APOSBIPG1132-1G
Azido-PEG10-amine
Azido-PEG10-amine
Formel: C₂₂H₄₆N₄O₁₀
Lagertemperatur: Raumtemperatur
MDL: MFCD03453246
CAS-Nummer: 912849-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR