Azido-PEG4-PFP ester

Lieferant: Apollo Scientific

BIPG1164-100MG
APOSBIPG1164-100MGEA 545 EUR
APOSBIPG1164-100MG
Azido-PEG4-PFP ester
Azido-PEG4-PFP ester
Formel: C₁₇H₂₀F₅N₃O₆
Molecular Weight: 457,349 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20926373
CAS-Nummer: 1353012-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR