Biotin-PEG2-Amine

Lieferant: Apollo Scientific
Achtung

BIPG1207-100MG BIPG1207-50MG BIPG1207-250MG
APOSBIPG1207-100MGEA 217 EUR
APOSBIPG1207-100MG APOSBIPG1207-50MG APOSBIPG1207-250MG
Biotin-PEG2-Amine
Biotin-PEG2-Amine
Formel: C₁₆H₃₀N₄O₄S
Lagertemperatur: Kühlschrank
MDL: MFCD09264745
CAS-Nummer: 138529-46-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR