Bis-PEG4-acid

Lieferant: Apollo Scientific
Achtung

BIPG1249-250MG
APOSBIPG1249-250MGEA 0 EUR
APOSBIPG1249-250MG
Bis-PEG4-acid
Bis-PEG4-acid
Formel: C₁₂H₂₂O₈
Lagertemperatur: Raumtemperatur
MDL: MFCD22201545
CAS-Nummer: 31127-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR