Bromoacetamido-PEG4- NHS ester

Lieferant: Apollo Scientific

BIPG1317-50MG
APOSBIPG1317-50MGEA 157 EUR
APOSBIPG1317-50MG
Bromoacetamido-PEG4- NHS ester
Bromoacetamido-PEG4- NHS ester
Formel: C₁₇H₂₇BrN₂O₉
Molecular Weight: 483,308 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20229159
CAS-Nummer: 1260139-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR