Fmoc-N-amido-PEG9-acid

Lieferant: Apollo Scientific

Synonyme: Fmoc-NH-PEG9-CH2CH2COOH, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28,31-decaoxa-4-azatetratriacontan-34-oic acid

BIPG1403-100MG
APOSBIPG1403-100MGEA 30.6 EUR
APOSBIPG1403-100MG
Fmoc-N-amido-PEG9-acid
Fmoc-N-amido-PEG9-acid
Formel: C₃₆H₅₃NO₁₃
Molecular Weight: 707.805 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28385470
CAS-Nummer: 1191064-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR