Fmoc-NH-PEG4-CH2CO2H

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-NH-PEG4-CH2COOH, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16-pentaoxa-4-azaoctadecan-18-oic acid

BIPG1410-500MG
APOSBIPG1410-500MGEA 522 EUR
APOSBIPG1410-500MG
Fmoc-NH-PEG4-CH2CO2H
Fmoc-NH-PEG4-CH2CO2H
Formel: C₂₅H₃₁NO₈
Molecular Weight: 473,516 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26142981
CAS-Nummer: 437655-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR