Fmoc-NH-PEG8-CH2COOH

Lieferant: Apollo Scientific
Achtung

BIPG1413-500MG
APOSBIPG1413-500MGEA 795 EUR
APOSBIPG1413-500MG
Fmoc-NH-PEG8-CH2COOH
Fmoc-NH-PEG8-CH2COOH
Formel: C₃₃H₄₇NO₁₂
Molecular Weight: 649,726 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27635163
CAS-Nummer: 868594-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR