Hydroxy-PEG3-acid

Lieferant: Apollo Scientific

BIPG1429-250MG BIPG1429-1G
APOSBIPG1429-250MGEA 0 EUR
APOSBIPG1429-250MG APOSBIPG1429-1G
Hydroxy-PEG3-acid
Hydroxy-PEG3-acid
Formel: C₉H₁₈O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD26793743
CAS-Nummer: 518044-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR