Hydroxy-PEG4-acid

Lieferant: Apollo Scientific
Achtung

BIPG1430-250MG BIPG1430-500MG BIPG1430-1G
APOSBIPG1430-250MGEA 0 EUR
APOSBIPG1430-250MG APOSBIPG1430-500MG APOSBIPG1430-1G
Hydroxy-PEG4-acid
Hydroxy-PEG4-acid
Formel: C₁₁H₂₂O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD22574805
CAS-Nummer: 937188-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR