Hydroxy-PEG2-CH2CO2H

Lieferant: Apollo Scientific
Achtung

BIPG1434-1G
APOSBIPG1434-1GEA 243 EUR
APOSBIPG1434-1G
Hydroxy-PEG2-CH2CO2H
Hydroxy-PEG2-CH2CO2H
Formel: C₆H₁₂O₅
Molecular Weight: 164,156 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20640451
CAS-Nummer: 51951-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR