Hydroxy-PEG1-CH2CO2tBu
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 2-(2-hydroxyethoxy)acetate, 2-Methyl-2-propanyl (2-hydroxyethoxy)acetate, Hydroxy-PEG1-CH2-Boc
BIPG1435-250MG
APOSBIPG1435-250MGEA
21.9
EUR
APOSBIPG1435-250MG
Hydroxy-PEG1-CH2CO2tBu
Hydroxy-PEG1-CH2CO2tBu
Formel:
C₈H₁₆O₄ Molecular Weight: 176.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27635155 CAS-Nummer: 287174-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...