Hydroxy-PEG1-t-butyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Hydroxy-PEG1-(CH2)2-Boc, tert-Butyl 3-(2-hydroxyethoxy)propanoate

BIPG1447-1G BIPG1447-500MG BIPG1447-5G
APOSBIPG1447-1GEA 69.6 EUR
APOSBIPG1447-1G APOSBIPG1447-500MG APOSBIPG1447-5G
Hydroxy-PEG1-t-butyl ester
Hydroxy-PEG1-t-butyl ester
Formel: C₉H₁₈O₄
Molecular Weight: 190.237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22056310
CAS-Nummer: 671802-00-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR