Hydroxy-PEG-5-t-butyl ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Hydroxy-PEG5-Boc, tert-Butyl 1-hydroxy-3,6,9,12,15-pentaoxaoctadecan-18-oate
BIPG1451-500MG
BIPG1451-250MG
BIPG1451-1G
APOSBIPG1451-500MGEA
219
EUR
APOSBIPG1451-500MG
APOSBIPG1451-250MG
APOSBIPG1451-1G
Hydroxy-PEG-5-t-butyl ester
Hydroxy-PEG-5-t-butyl ester
Formel:
C₁₇H₃₄O₈ Dichte: 1,071 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD20926402 CAS-Nummer: 850090-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...