Mal-PEG3-acid

Lieferant: Apollo Scientific

Synonyme: 3-(2-(2-(2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy)ethoxy)ethoxy)propanoic acid, Maleimido-tri(ethylene glycol)-propionic acid

BIPG1493-100MG
APOSBIPG1493-100MGEA 69.6 EUR
APOSBIPG1493-100MG
Mal-PEG3-acid
Mal-PEG3-acid
Formel: C₁₃H₁₉NO₇
Molecular Weight: 301.292 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23103943
CAS-Nummer: 518044-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR