Methyl-PEG12-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 2,5,8,11,14,17,20,23,26,29,32,35-Dodecaoxaoctatriacontan-38-oic acid, m-PEG12-acid

BIPG1551-100MG BIPG1551-50MG
APOSBIPG1551-100MGEA 473 EUR
APOSBIPG1551-100MG APOSBIPG1551-50MG
Methyl-PEG12-carboxylic acid
Methyl-PEG12-carboxylic acid
Formel: C₂₆H₅₂O₁₄
Molecular Weight: 588.69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08274611
CAS-Nummer: 2135793-73-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR