Octaethylene glycol monomethyl ether

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5,8,11,14,17,20,23-Octaoxapentacosan-25-ol, mPEG8-Alcohol

BIPG1563-1G
APOSBIPG1563-1GEA 294 EUR
APOSBIPG1563-1G
Octaethylene glycol monomethyl ether
Octaethylene glycol monomethyl ether
Formel: C₁₇H₃₆O₉
Molecular Weight: 384,47 g/mol
Siedepunkt: 216 °C
Dichte: 1,09 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD06797152
CAS-Nummer: 25990-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR