Decaethylene glycol monomethyl ether

Lieferant: Apollo Scientific
Achtung

BIPG1565-1G
APOSBIPG1565-1GEA 0 EUR
APOSBIPG1565-1G
Decaethylene glycol monomethyl ether
Decaethylene glycol monomethyl ether
Formel: C₂₁H₄₄O₁₁
Lagertemperatur: Raumtemperatur
MDL: MFCD06797087
CAS-Nummer: 27425-92-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR