m-PEG3-Amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-(2-Methoxyethoxy)ethoxy)ethanamine

BIPG1581-500MG
APOSBIPG1581-500MGEA 522 EUR
APOSBIPG1581-500MG
m-PEG3-Amine
m-PEG3-Amine
Formel: C₇H₁₇NO₃
Molecular Weight: 163,215 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17215909
CAS-Nummer: 74654-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR