2-(2-Propynyloxy)ethylamine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Propargyl-PEG1-amine, Propargyl-PEG1-NH2
BIPG1722-100MG
BIPG1722-250MG
APOSBIPG1722-100MGEA
27.7
EUR
APOSBIPG1722-100MG
APOSBIPG1722-250MG
2-(2-Propynyloxy)ethylamine
2-(2-Propynyloxy)ethylamine
Formel:
C₅H₉NO Molecular Weight: 99.13 g/mol Dichte: 0.96 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD16713446 CAS-Nummer: 122116-12-5 UN: 2734 ADR: 8+3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...