t-Boc-N-amido-PEG4-acid

Lieferant: Apollo Scientific
Achtung

BIPG1802-500MG BIPG1802-100MG BIPG1802-1G
APOSBIPG1802-500MGEA 277 EUR
APOSBIPG1802-500MG APOSBIPG1802-100MG APOSBIPG1802-1G
t-Boc-N-amido-PEG4-acid
t-Boc-N-amido-PEG4-acid
Formel: C₁₆H₃₁NO₈
Lagertemperatur: Kühlschrank
CAS-Nummer: 756525-91-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR