21-[(tert-Butoxycarbonyl)amino]-4,7,10,13,16,19-hexaoxaheneicosanoic acid

Lieferant: Apollo Scientific
Achtung

BIPG1804-100MG
APOSBIPG1804-100MGEA 81.2 EUR
APOSBIPG1804-100MG
21-[(tert-Butoxycarbonyl)amino]-4,7,10,13,16,19-hexaoxaheneicosanoic acid
21-[(tert-Butoxycarbonyl)amino]-4,7,10,13,16,19-hexaoxaheneicosanoic acid
Formel: C₂₀H₃₉NO₁₀
Lagertemperatur: Kühlschrank
MDL: MFCD07781255
CAS-Nummer: 882847-13-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR