t-Boc-N-Amido-PEG5-Amine

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl (17-amino-3,6,9,12,15-pentaoxaheptadecyl)carbamate

BIPG1813-100MG BIPG1813-250MG BIPG1813-500MG
APOSBIPG1813-100MGEA 79.9 EUR
APOSBIPG1813-100MG APOSBIPG1813-250MG APOSBIPG1813-500MG
t-Boc-N-Amido-PEG5-Amine
t-Boc-N-Amido-PEG5-Amine
Formel: C₁₇H₃₆N₂O₇
Lagertemperatur: Kühlschrank
MDL: MFCD22056313
CAS-Nummer: 189209-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR