t-Boc-N-Amido-PEG7-Amine

Lieferant: Apollo Scientific
Achtung

BIPG1815-100MG BIPG1815-50MG BIPG1815-250MG
APOSBIPG1815-100MGEA 122 EUR
APOSBIPG1815-100MG APOSBIPG1815-50MG APOSBIPG1815-250MG
t-Boc-N-Amido-PEG7-Amine
t-Boc-N-Amido-PEG7-Amine
Formel: C₂₁H₄₄N₂O₉
Lagertemperatur: Kühlschrank
MDL: MFCD03453229
CAS-Nummer: 206265-98-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR