t-Boc-N-Amido-PEG2-CH2CO2H

Lieferant: Apollo Scientific

Synonyme: Boc-NH-PEG2-CH2COOH, 2,2-Dimethyl-4-oxo-3,8,11-trioxa-5-azatridecan-13-oic acid

BIPG1825-1G
APOSBIPG1825-1GEA 29 EUR
APOSBIPG1825-1G
t-Boc-N-Amido-PEG2-CH2CO2H
t-Boc-N-Amido-PEG2-CH2CO2H
Formel: C₁₁H₂₁NO₆
Molecular Weight: 263.288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17019374
CAS-Nummer: 108466-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR