Thiol-PEG4-acid

Lieferant: Apollo Scientific
Achtung

BIPG1838-250MG
APOSBIPG1838-250MGEA 270 EUR
APOSBIPG1838-250MG
Thiol-PEG4-acid
Thiol-PEG4-acid
Formel: C₁₁H₂₂O₆S
Lagertemperatur: Raumtemperatur
CAS-Nummer: 749247-06-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR