Thiol-PEG6-acid

Lieferant: Apollo Scientific

BIPG1839-250MG
APOSBIPG1839-250MGEA 788 EUR
APOSBIPG1839-250MG
Thiol-PEG6-acid
Thiol-PEG6-acid
Formel: C₁₅H₃₀O₈S
MDL: MFCD26127814
CAS-Nummer: 1347750-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR