Thiol-PEG3-Alcohol

Lieferant: Apollo Scientific

BIPG1840-1G
APOSBIPG1840-1GEA 935 EUR
APOSBIPG1840-1G
Thiol-PEG3-Alcohol
Thiol-PEG3-Alcohol
Formel: C₆H₁₄O₃S
Molecular Weight: 166,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116140
CAS-Nummer: 56282-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR