Gellan Gummi, powder

Lieferant: Apollo Scientific
Achtung

BIPG434-100G BIPG434-1KG BIPG434-500G
APOSBIPG434-100GEA 94.4 EUR
APOSBIPG434-100G APOSBIPG434-1KG APOSBIPG434-500G
Gellan Gummi, powder
Gellan Gummi
Schmelzpunkt: >70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00131909
CAS-Nummer: 71010-52-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR