Recombinant collagenase G
Lieferant: Apollo Scientific
BIPR1004-2000C
APOSBIPR1004-2000CEA 220 EUR
APOSBIPR1004-2000C
Recombinant collagenase G
Enzyme
From Clostridium histolyticum and produced in E. Coli and B. Megaterium. Endotoxin negative. No detectable Caseinase activity. Sterile lyophilized powder.