Rifampicin 99% approx

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,6,9,17,19,21-Hexahydroxy-23-methoxy- 2,4,12,16,18,20,22-heptamethyl-8-[N-(4-methyl- 1-piperazinyl)formimidoyl]-2,7-(epoxypentadeca- [1,11,13]trienimino)-naphtho[2,1-b]furan- 1,11(2H)-dion-21-acetat, Rifampin

BIR0146-1G BIR0146-25G BIR0146-5G
APOSBIR0146-1GEA 21.9 EUR
APOSBIR0146-1G APOSBIR0146-25G APOSBIR0146-5G
Rifampicin 99% approx
Rifampicin

Rifampicin is active against gram-positive but less active against gram-negative bacteria.


  • Cell culture tested

Formel: C₄₃H₅₈N₄O₁₂
Schmelzpunkt: 183…194 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00151389
CAS-Nummer: 13292-46-1
EINECS: 236-312-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR