Fast Red RC Salt

Lieferant: Apollo Scientific
Achtung

BIR4011-5G
APOSBIR4011-5GEA 21.9 EUR
APOSBIR4011-5G
Fast Red RC Salt
Fast Red RC Salt
Formel: C₇H₆Cl₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00074764
CAS-Nummer: 68025-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR