Sulphamethoxazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-N-(5-methylisoxazol-3-yl)benzenesulphonamide
BIS0149-25G
APOSBIS0149-25GEA
21.9
EUR
APOSBIS0149-25G
Sulphamethoxazole
Sulphamethoxazole
- Antibodies
Formel:
C₁₀H₁₁N₃O₃S Molecular Weight: 253,28 g/mol Schmelzpunkt: 166…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010546 CAS-Nummer: 723-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...