Sulbactam sodium

Lieferant: Apollo Scientific
Achtung

Synonyme: Sodium (2S-cis)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4,4-dioxide

BIS0150-10G BIS0150-1G BIS0150-5G
APOSBIS0150-10GEA 386 EUR
APOSBIS0150-10G APOSBIS0150-1G APOSBIS0150-5G
Sulbactam sodium
Sulbactam sodium
Formel: C₈H₁₀NO₅S·Na
Molecular Weight: 255.223 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01750374
CAS-Nummer: 69388-84-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR