1,9-Pyrazoloanthrone ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125
BIS1100-1G
BIS1100-300MG
APOSBIS1100-1GEA
444
EUR
APOSBIS1100-1G
APOSBIS1100-300MG
1,9-Pyrazoloanthrone ≥99%
1,9-Pyrazoloanthrone
Novel, potent and selective JNK-1,-2, and -3 inhibitor (Ki = 0.19 uM). SP600125 is an ATP-competitive reversible inhibitor with >20-fold selectivity vs. a range of kinases and enzymes tested. Proc. Natl. Acad. Sci. USA 98: 13681-13686 (2001).
Formel:
C₁₄H₈N₂O Molecular Weight: 220.231 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00022289 CAS-Nummer: 129-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...