Sterigmatocystin
Lieferant: Apollo Scientific
|
Gefahr
|
BIS1401-1MG
APOSBIS1401-1MGEA
46.4
EUR
APOSBIS1401-1MG
Sterigmatocystin
Sterigmatocystin
Formel:
C₁₈H₁₂O₆ Lagertemperatur: Gefrierschrank |
CAS-Nummer:
10048-13-2 EINECS: 233-158-6 UN: 3462 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...