D(+)-Saccharose

Lieferant: Apollo Scientific
Achtung

Synonyme: Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker

BIS3391-1KG BIS3391-500G
APOSBIS3391-1KGEA 27.7 EUR
APOSBIS3391-1KG APOSBIS3391-500G
D(+)-Saccharose
D(+)-Saccharose

  • Plant tissue culture tested

Formel: C₁₂H₂₂O₁₁
Molecular Weight: 342.3 g/mol
Schmelzpunkt: 185…190 °C
Dichte: 1.58 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006626
CAS-Nummer: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR