Streptozocin
Lieferant: Apollo Scientific
|
Gefahr
|
BIS4356-1G
BIS4356-250MG
BIS4356-500MG
APOSBIS4356-1GEA
702
EUR
APOSBIS4356-1G
APOSBIS4356-250MG
APOSBIS4356-500MG
Streptozocin
Streptozocin
Potent methylating agent for DNA
Formel:
C₈H₁₅N₃O₇ Schmelzpunkt: 121 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00006607 CAS-Nummer: 18883-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...